Skip to Content
Merck

BDPH9C021C19

Bide Pharmatech

(E)-3-(2-Propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid

Synonym(s):

(E)-3-(2-propyl-6-(trifluoromethyl)pyridin-3-yl)acrylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C12H12O2N1F3
CAS Number:
Molecular Weight:
259.23
MDL number:
Product BDPH9C021C19 is not currently sold in your country.


Inchi Code

1S/C12H12F3NO2/c1-2-3-9-8(5-7-11(17)18)4-6-10(16-9)12(13,14)15/h4-7H,2-3H2,1H3,(H,17,18)/b7-5+

Inchi Key

CNCIJHJOEHBPBF-FNORWQNLSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library