Skip to Content
Merck

ENAH31551B58

Enamine

2-amino-4H,5H,6H-cyclopenta[b]thiophene-3-carboxylic acid hydrochloride

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENAH31551B58 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
219.69
MDL number:
Product ENAH31551B58 is not currently sold in your country.

Iupac Name

2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylic acid hydrochloride

Inchi Code

1S/C8H9NO2S.ClH/c9-7-6(8(10)11)4-2-1-3-5(4)12-7;/h1-3,9H2,(H,10,11);1H

InChI key

SUGWLOAEXHFNDR-UHFFFAOYSA-N

storage temp.

RT

Salt Data

Cl

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-6731069

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.enaminestore.com/catalog/EN300-6731069


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library