Skip to Content
Merck

ABC441

Anti-ZKSCAN3 (Zf47)

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein with KRAB and SCAN domains 3, Zinc finger and SCAN domain-containing protein 13, Zinc finger protein 306, Zinc finger protein 309, Zinc finger protein 47 homolog, Zf47, Zfp-47

Sign In to View Organizational & Contract Pricing

Select a Size

25 μL

$284.00

100 μL

$672.00

$284.00


Check Cart for Availability

Request a Bulk Order

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... ZKSCAN3(80317)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
ABE1341SAB4503345AB4340
antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Gene Information

human ... ZKSCAN3(80317)

Gene Information

human ... ZBTB38(253461)

Gene Information

human ... ZNF397(84307)

Gene Information

-

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunohistochemistry: suitable (paraffin), western blot: suitable

technique(s)

ELISA: 1:5000, western blot: 1:500-1:1000

technique(s)

immunoprecipitation (IP): suitable, western blot: suitable

General description

Zinc finger protein with KRAB and SCAN domains 3 (UniProt: Q9BRR0; also known as Zinc finger and SCAN domain-containing protein 13, Zinc finger protein 306, Zinc finger protein 309, Zinc finger protein 47 homolog, Zf47, Zfp-47) is encoded by the ZKSCAN3 (also known as ZFP47, ZNF306, ZNF309, ZSCAN13) gene (Gene ID: 80317) in human. Zf47 is a member of the krueppel C2H2-type zinc-finger protein family. Two isoforms of Zf47 have been described that are produced by alternative splicing. It contains 7 C2H2 type zinc-fingers. It serves as a transcriptional factor that binds to the consensus sequence 5′-[GT][AG][AGT]GGGG-3′ and acts as a repressor of autophagy. It is shown to specifically repress expression of genes involved in autophagy and lysosome biogenesis/function such as MAP1LC3B, ULK1 or WIPI2. It is mainly localized in the nucleus, however, under conditions of starvation, it translocates to the cytoplasm, allowing expression of target genes involved in autophagy and lysosome biogenesis/function. Overexpression of Zf47 is observed in various tumors, including multiple myeloma, colorectal and prostate cancers.
~65 kDa observed; 60.64 kDa calculated. Uncharacterized bands may be observed in some lysate(s).

Immunogen

Epitope: unknown
KLH-conjugated linear peptide corresponding to 12 amino acids from the N-terminal half of human Zinc finger protein with KRAB and SCAN domains 3. The immunogen sequence is conserved in both isoforms, 1 and 2.

Application

Anti-ZKSCAN3 (Zf47), Cat. No. ABC441, is a rabbit polyclonal antibody that detects Zinc finger protein with KRAB and SCAN domains 3 and has been tested for use in Immunocytochemistry and Western Blotting.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ZKSCAN3 (Zf47) with KRAB and SCAN domains 3 in HeLa and A431 cell lines.
Research Category
Apoptosis & Cancer

Biochem/physiol Actions

This rabbit polyclonal antibody detects Zinc finger protein with KRAB and SCAN domains 3 in human cells. It targets an epitope within 12 amino acids from the N-terminal half.

Physical form

Affinity Purified
Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ZKSCAN3 (Zf47) with KRAB and SCAN domains 3 in HeLa cell lysate.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service