Skip to Content
Merck

TU-10X5ML-U

Blood typing Unlabeled Vials Reagent - RHCE (Rh) Specificities

Anti-e from the clone MS-16/MS-21/MS-63, Human IgM, Blood Grouping Reagent

Synonym(s):

Anti-e (clone MS-16/MS-21/MS-63), Blood Grouping Reagent

Sign In to View Organizational & Contract Pricing

About This Item

UNSPSC Code:
41116102
NACRES:
NA.41

Note: For further information on price, availability or to place an order, please contact us

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

human

Quality Level

antibody form

unpurified

clone

MS-16/MS-21/MS-63, monoclonal

form

liquid

packaging

pkg of 10 × 5 mL

concentration

15-30 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

7.1-7.3

isotype

IgM

shipped in

ambient

storage temp.

2-8°C

Compare Similar Items

View Full Comparison

Show Differences

1 of 2

This Item
TA-10X5ML-U
antibody form

unpurified

antibody form

unpurified

biological source

human

biological source

human

clone

MS-16/MS-21/MS-63, monoclonal

clone

MS-80/MS-258, monoclonal

isotype

IgM

isotype

IgM

Quality Level

600

Quality Level

600

storage temp.

2-8°C

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 30 months from date of filtration

For the manufacture of blood typing IVD′s

For In Vitro use only

Application

For use by Tube;Slide;Microplate;Gel and Column techniques
Vialled, Unlabeled

Legal Information

Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service