Skip to Content
Merck

BDPH9BCD4792

Bide Pharmatech

(5-Methylpyrazin-2-yl)methanol

Synonym(s):

(5-methyl-2-pyrazinyl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O1N2
CAS Number:
Molecular Weight:
124.14
MDL number:
Product BDPH9BCD4792 is not currently sold in your country.


Inchi Code

1S/C6H8N2O/c1-5-2-8-6(4-9)3-7-5/h2-3,9H,4H2,1H3

Inchi Key

QYANNJBVADZUDN-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or Semi-Solid or Solid or Lump



Still not finding the right product?

Explore all of our products under (5-Methylpyrazin-2-yl)methanol


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library