Skip to Content
Merck

ABN192

Anti-ZFP36L1 Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

Zinc finger protein 36, C3H1 type-like 1, Butyrate response factor 1, EGF-response factor 1, ERF-1, Protein TIS11B

Sign In to View Organizational & Contract Pricing.

Select a Size

100 μG

€452.00

€452.00


Estimated to ship on26 December 2025Details


Request a Bulk Order

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-ZFP36L1 Antibody, from rabbit, purified by affinity chromatography

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human

species reactivity (predicted by homology)

bovine (based on 100% sequence homology), canine (based on 100% sequence homology), horse (based on 100% sequence homology), mouse (based on 100% sequence homology), rat (based on 100% sequence homology)

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Quality Level

Gene Information

human ... ZFP36L1(677)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
WH0000677M2AV38875HPA014909
antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

antibody form

IgG fraction of antiserum

antibody form

affinity isolated antibody

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

biological source

rabbit

biological source

mouse

biological source

rabbit

biological source

rabbit

technique(s)

western blot: suitable

technique(s)

indirect ELISA: suitable, western blot: 1-5 μg/mL

technique(s)

western blot: suitable

technique(s)

immunoblotting: 0.04-0.4 μg/mL, immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500

species reactivity

human

species reactivity

human

species reactivity

pig, human, rabbit, dog, rat, bovine, horse, mouse

species reactivity

human

Gene Information

human ... ZFP36L1(677)

Gene Information

human ... ZFP36L1(677)

Gene Information

human ... ZFP36L2(678)

Gene Information

human ... ZFPL1(7542)

Analysis Note

Control
HeLa cell lysate
Evaluated by Western Blot in HeLa cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected ZFP36L1 in 10 µg of HeLa cell lysate.

Application

Anti-ZFP36L1 detects levels of ZFP36L1 proteins & has been published & validated for use in WB.
Research Category
Neuroscience
Research Sub Category
Neurodegenerative Diseases

Biochem/physiol Actions

This antibody recognizes the C-terminus of ZFP36L1.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

General description

As a member of the ZFP36/tristetraprolin (TTP) family of zinc finger proteins, ZFP36L1 serves as an RNA-binding protein (RBP) that is known to interact with AU-rich elements in the 3’-untranslated region of mRNAs, resulting in their degradation. It serves as a target for Keratinocyte-derived growth factors and cytokines. ZFP36L1 is reported to play an important role in various physiological processes, including wound healing, epidermal homeostasis, inflammation, erythroid differentiation, thymocyte development, and in the prevention of malignant transformation. Defects in the ZFPP36L1 gene have been implicated in Mutiple Sclerosis and cancer.
~38 kDa observed

Immunogen

Epitope: C-terminus
KLH-conjugated linear peptide corresponding to the C-terminus of Human ZFP36L1.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Branka Popović et al.
Cell reports, 42(5), 112419-112419 (2023-04-19)
Potent T cell responses against infections and malignancies require a rapid yet tightly regulated production of toxic effector molecules. Their production level is defined by post-transcriptional events at 3' untranslated regions (3' UTRs). RNA binding proteins (RBPs) are key regulators in
Ming-Tai Chen et al.
Scientific reports, 5, 16229-16229 (2015-11-07)
RNA binding proteins (RBPs)-mediated post-transcriptional control has been implicated in influencing various aspects of RNA metabolism and playing important roles in mammalian development and pathological diseases. However, the functions of specific RBPs and the molecular mechanisms through which they act

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service