Skip to Content
Merck

MABS1284

Anti-Apolipoprotein(a) Antibody, clone LPA4

clone LPA4, from mouse

Synonym(s):

Apolipoprotein(a), Apo(a), Lp(a)

Sign In to View Organizational & Contract Pricing

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

LPA4, monoclonal

species reactivity

human, primate

technique(s)

ELISA: suitable
immunohistochemistry: suitable (paraffin)
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... APOA1(335)

General description

499.19/501.3 kDa (mature/pro-form) calculated based on the isoform sequence reported by UniProt (P08519). Note: There are over 40 isoforms of different sizes reported in literature with varying number of KIV2 repeats.
Apolipoprotein(a) (UniProt P08519; also known as Apo(a)) is encoded by the LPA gene (Gene ID 4018) in human. Lipoprotein (a)/Lp(a) is composed of apolipoprotein (a)/apo(a) covalently bound to apolipoprotein B-100/apoB via a single disulfide bond on kringle (K)IV type 9 (KIV9) to a site near apoB low density lipoprotein (LDL) receptor-binding site. The LPA gene that encodes Apo(a) appeared late during primate evolution and is not present in non-primate species. Genetically elevated Lp(a) level is a risk factor for cardiovascular disease (CVD) and peripheral arterial disease. Lp(a) is a preferential lipoprotein carrier of oxidized phospholipids (OxPLs) and circulating levels of oxidized LDL are strongly associated with angiographically documented coronary artery disease (CAD). Structurally, Ap(a) contains ten subtypes of KIV sequences, with the KIV2 subtype present in multiple identical repeats, followed by a KV sequence and an inactive protease domain. There are over 40 isoforms of different sizes reported in literature with varying number of KIV2 repeats. The KIV9 contains the cysteine that mediates covalent linkage of Apo(a) to ApoB, while the inactive protease domain competes against plasminogen (PLG) activators (PAs) for PLG binding, thereby preventing the conversion of PLG to plasmin.

Immunogen

Human lipoprotein(a)/Lp(a) (Tsimikas, S., et al. (2004). Circulation. 109(25):3164-3170).

Application

Detect Apolipoprotein(a)/Apo(a)/Lp(a) using this mouse monoclonal Anti-Apolipoprotein(a) Antibody, clone LPA4, Cat. No. MABS1284, validated for use in and ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, and Western Blotting.
ELISA Analysis: A representative lot detected patients plasma Lp(a) levels (5 to 175 mg/dL) as both the capture (5 µg/mL, 50 µL/well; unconjugated) and the detection (1 µg/mL; biotinconjugated) antibody by sandwich ELISA (Courtesy of Dr. Sotirios Tsimikas, M.D., University of California at San Diego, USA).

Immunohistochemistry Analysis: A 1:25 dilution from a representative lot (pre-conjugated with biotin) detected Apo(a) in paraffin-embedded human carotid artery tissue sections (Courtesy of Dr. Sotirios Tsimikas, M.D., University of California at San Diego, USA).

ELISA Analysis: A representative lot was employed as either the capture or detection antibody for the detection of Lp(a) levels in human, chimpanzee, bonobo, gorilla, baboon, and cynomolgus monkey plasma samples, as well as the interaction of OxPL and ApoB with various human Apo(a) transgenes in Tg mice blood samples and purified recombinant human Apo(a) constructs (Leibundgut, G., et al. (2013). J. Lipid Res. 54(10):2815-2830).

ELISA Analysis: A representative lot was employed either as the capture antibody for the detection of Apo(a) and Apo(a)-associated OxPL or as the detection antibody (biotinylated) for the detection of Lp(a) (ApoB-linked Apo(a)) captured by an ApoB antibody in a atherosclerosis study involving uremic mice that express human Apo(a) and ApoB transgene (Pedersen, T.X., et al. (2010). J. Lipid Res. 51(10):2967-2975).

Immunoprecipitation Analysis: A representative lot immunodepleted Apo(a) and associated OxLDL-E06 from patients plasma samples collected immediately before or 6 hrs after percutaneous coronary intervention (PCI), while only ~50% of OxLDL-E06 was found associated and co-precipitated with Apo(a) in the immediate post-PCI time point (Tsimikas, S., et al. (2004). Circulation. 109(25):3164-3170).

Western Blotting Analysis: A representative lot detected recombinant human wild-type Apo(a) and a 8K-IV lysine-binding defective (LBS-deficient) Apo(a) mutant expressed in and purified from transfected HEK293 cells (Leibundgut, G., et al. (2013). J. Lipid Res. 54(10):2815-2830).
Research Category
Signaling

Biochem/physiol Actions

Clone LPA4 targets a 14-a.a. sequence present in human apolipoprotein(a)/Apo(a) KIV-5, KIV-7, and KIV-8 (Leibundgut, G., et al. (2013). J. Lipid Res. 54(10):2815-2830), or Kringle domain 32, 34, and 45 based on the isoform sequence reported by UniProt (P08519), making it possible to use this clone as both the capture (unconjugated) and detection (conjugated) antibody in sandwich ELISA application for the detection of Apo(a) and Lp(a) (ApoB-linked Apo(a)). Clone LPA4 does not cross-react with the highly homologous plasminogen. Note that the target detected by clone LPA4 is not lysophosphatidic acid receptor 4 (a.k.a. LPA-4; UniProt Q99677) encoded by the LPAR4 (a.k.a. LPA4) gene, nor apolipoproteins A-I, A-II, A-IV, and A-V encoded by the APOA1, APOA2, APOA4, APOA5 genes.

Physical form

Format: Purified
Protein G purified.
Purified mouse IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Identity Confirmation by Isotyping Test.

Isotyping Analysis: The identity of this monoclonal antibody is confirmed by isotyping test to be mouse IgG1 .

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Maximillian A Rogers et al.
Frontiers in cardiovascular medicine, 9, 778919-778919 (2022-02-15)
Lipoprotein(a) (Lp[a]) blood levels >50 mg/dL is a major cardiovascular disease risk factor in humans. Lp(a) associates with increased cardiovascular calcification, a critical pathology with no clinically available drug therapies. The mechanisms through which Lp(a) increases cardiovascular calcification risk remain
Kazuhiro Takara et al.
Cell reports, 20(9), 2072-2086 (2017-08-31)
Vascular normalization in tumors may improve drug delivery and anti-tumor immunity. Angiogenesis inhibitors induce hypoxia, which may facilitate malignant progression; therefore, we investigated other methods to promote vascular maturation. Here, we show that lysophosphatidic acid (LPA) enhances blood flow by
Gregory T Jones et al.
Epigenetics, 15(9), 949-958 (2020-04-03)
Changes in whole blood DNA methylation levels at several CpG sites have been associated with circulating blood lipids, specifically high-density lipoprotein and triglycerides. This study performs a discovery and validation epigenome-wide association study (EWAS) for circulating lipoprotein(a) [Lp(a)], an independent
Raphaëlle Bourgeois et al.
JACC. Basic to translational science, 5(9), 888-897 (2020-10-06)
Our objectives were to determine whether autotaxin (ATX) is transported by lipoprotein(a) [Lp(a)] in human plasma and if could be used as a biomarker of calcific aortic valve stenosis (CAVS). We first found that ATX activity was higher in Lp(a)
Full Issue PDF.
JACC. Basic to translational science, 5(9), I-CV (2020-10-06)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service