Skip to Content
Merck

53200

Hexyllithium solution

purum, ~2.3 M in hexane

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Linear Formula:
CH3(CH2)5Li
CAS Number:
Molecular Weight:
92.11
Beilstein:
3600261
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

grade

purum

concentration

~2.3 M in hexane

color

yellow-brown

density

0.70 g/mL at 20 °C

storage temp.

2-8°C

SMILES string

[Li]CCCCCC

InChI

1S/C6H13.Li/c1-3-5-6-4-2;/h1,3-6H2,2H3;

InChI key

RMKNFFPDYCVCDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Strong base; no gaseous by-product is formed upon workup

comparable product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Pyr. Liq. 1 - Repr. 2 - Skin Corr. 1A - STOT RE 2 Inhalation - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Nervous system

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

15.8 °F - closed cup

Flash Point(C)

-9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service