Skip to Content
Merck

SAB4700408

Monoclonal Anti-CD18-PE antibody produced in mouse

clone MEM-48, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ITGB2, Anti-Integrin β2 subunit

Sign In to View Organizational & Contract Pricing

Select a Size

100 TEST

€422.00

€422.00


Please contact Customer Service for Availability


About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

mouse

conjugate

phycoerythrin (R-PE) conjugate

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MEM-48, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

flow cytometry: suitable

isotype

IgG1

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ITGB2(3689)

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
SAB4700088SAB4700113SAB4700196
species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

nonhuman primates, human

biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

antibody form

purified immunoglobulin

clone

MEM-48, monoclonal

clone

MEM-31, monoclonal

clone

LT19, monoclonal

clone

MEM-102, monoclonal

conjugate

phycoerythrin (R-PE) conjugate

conjugate

phycoerythrin (R-PE) conjugate

conjugate

phycoerythrin (R-PE) conjugate

conjugate

phycoerythrin (R-PE) conjugate

Gene Information

human ... ITGB2(3689)

Gene Information

human ... CD8(925)

Gene Information

human ... CD19(930)

Gene Information

human ... CD48(962)

General description

The antibody MEM-48 recognizes an epitope involving residues 534-546 in cysteine-rich repeat 3 of the CD18 antigen (integrin beta2 subunit; beta2 integrin). CD18 is a 90-95 kDa type I transmembrane protein expressed on all leukocytes.

Immunogen

Leukocytes of a patient suffering from a LGL-type leukemia

Application

The reagent is designed for Flow Cytometry analysis of human blood cells using 20 μL reagent / 100 μL of whole blood or 1e6 cells in a suspension. The content of a vial (2 mL) is sufficient for 100 tests.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline containing 15 mM sodium azide and 0.2% high-grade protease free BSA as a stabilizing agent.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service