Skip to Content
Merck

SML3081

AG14361

≥98% (HPLC)

Synonym(s):

2-[4-[(Dimethylamino)methyl]phenyl]-5,6-dihydroimidazo[4,5,1-jk][1,4]benzodiazepin-7(4H)-one, AG 14361, AG-14361

Sign Into View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
C19H20N4O
CAS Number:
Molecular Weight:
320.39
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C1C2=C3C(N=C(N3CCN1)C4=CC=C(C=C4)CN(C)C)=CC=C2

InChI

1S/C19H20N4O/c1-22(2)12-13-6-8-14(9-7-13)18-21-16-5-3-4-15-17(16)23(18)11-10-20-19(15)24/h3-9H,10-12H2,1-2H3,(H,20,24)

InChI key

SEKJSSBJKFLZIT-UHFFFAOYSA-N

Biochem/physiol Actions

AG14361 is a potent oly(ADP-ribose) polymerase-1 inhibitor (human PARP-1 Ki = 5.8 nM) that suppresses 25 μM MNNG-induced PARP-1 activity (NAD+ depletion = 70% without vs.12% with 0.4 μM AG14361; 87% reduced ADP-ribose polymer induction with 0.4 μM AG14361) and greatly amplifies temozolomide and topotecan cytotoxicity (2.2- and 2.0-fold lower IG50, respectively, with 0.4 μM AG14361) in A549 cultures. AG14361 is shown to selectively kill BRCA2-deficient cells over BRCA2-expressing cells both in cultures (1-10 μM) and in mice in vivo (25 mg/10 mL/kg/d ip).
Potent poly(ADP-ribose) polymerase-1 (PARP-1) inhibitor in vitro and in vivo.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Donald J Skalitzky et al.
Journal of medicinal chemistry, 46(2), 210-213 (2003-01-10)
Novel tricyclic benzimidazole carboxamide poly(ADP-ribose) polymerase-1 (PARP-1) inhibitors have been synthesized. Several compounds were found to be powerful chemopotentiators of temozolomide and topotecan in both A549 and LoVo cell lines. In vitro inhibition of PARP-1 was confirmed by direct measurement
Qiyang Shou et al.
JCI insight, 4(12) (2019-06-21)
The activation and recruitment of NK cells to the site of viral infection are crucial for virus control. However, it remains largely unknown what controls the recruitment of the activated NK cells to the infection site. In a model of
Helen E Bryant et al.
Nature, 434(7035), 913-917 (2005-04-15)
Poly(ADP-ribose) polymerase (PARP1) facilitates DNA repair by binding to DNA breaks and attracting DNA repair proteins to the site of damage. Nevertheless, PARP1-/- mice are viable, fertile and do not develop early onset tumours. Here, we show that PARP inhibitors
Aashish Soni et al.
Molecular cancer therapeutics, 17(10), 2206-2216 (2018-07-05)
Parp inhibitors (Parpi) are commonly used as single agents for the management of tumors with homologous recombination repair (HRR) deficiencies, but combination with radiotherapy (RT) is not widely considered due to the modest radiosensitization typically observed. BMN673 is one of
Sandra M Martín-Guerrero et al.
PloS one, 12(10), e0187130-e0187130 (2017-10-27)
Poly(ADP-ribose)polymerases (PARPs) are a family of NAD+ consuming enzymes that play a crucial role in many cellular processes, most clearly in maintaining genome integrity. Here, we present an extensive analysis of the alteration of mitochondrial morphology and the relationship to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service