Merck
  • TGF-β1, but not bone morphogenetic proteins, activates Smad1/5 pathway in primary human macrophages and induces expression of proatherogenic genes.
TGF-β1, but not bone morphogenetic proteins, activates Smad1/5 pathway in primary human macrophages and induces expression of proatherogenic genes.
Journal of immunology (Baltimore, Md. : 1950) (2014-12-17)
Dinara Nurgazieva, Amanda Mickley, Kondaiah Moganti, Wen Ming, Illya Ovsyi, Anna Popova, Sachindra, Kareem Awad, Nan Wang, Karen Bieback, Sergij Goerdt, Julia Kzhyshkowska, Alexei Gratchev
ABSTRACT
Macrophages are responsible for the control of inflammation and healing, and their malfunction results in cardiometabolic disorders. TGF-β is a pleiotropic growth factor with dual (protective and detrimental) roles in atherogenesis. We have previously shown that in human macrophages, TGF-β1 activates Smad2/3 signaling and induces a complex gene expression program. However, activated genes were not limited to known Smad2/3-dependent ones, which prompted us to study TGF-β1-induced signaling in macrophages in detail. Analysis of Id3 regulatory sequences revealed a novel enhancer, located between +4517 and 4662 bp, but the luciferase reporter assay demonstrated that this enhancer is not Smad2/3 dependent. Because Id3 expression is regulated by Smad1/5 in endothelial cells, we analyzed activation of Smad1/5 in macrophages. We demonstrate here for the first time, to our knowledge, that TGF-β1, but not BMPs, activates Smad1/5 in macrophages. We show that an ALK5/ALK1 heterodimer is responsible for the induction of Smad1/5 signaling by TGF-β1 in mature human macrophages. Activation of Smad1/5 by TGF-β1 induces not only Id3, but also HAMP and PLAUR, which contribute to atherosclerotic plaque vulnerability. We suggest that the balance between Smad1/5- and Smad2/3-dependent signaling defines the outcome of the effect of TGF-β on atherosclerosis where Smad1/5 is responsible for proatherogenic effects, whereas Smad2/3 regulate atheroprotective effects of TGF-β.
Product Description
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium Fluoride Solution
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder