Merck
  • Repeated remote ischemic conditioning attenuates left ventricular remodeling via exosome-mediated intercellular communication on chronic heart failure after myocardial infarction.
Repeated remote ischemic conditioning attenuates left ventricular remodeling via exosome-mediated intercellular communication on chronic heart failure after myocardial infarction.
International journal of cardiology (2014-12-03)
Takehiro Yamaguchi, Yasukatsu Izumi, Yasuhiro Nakamura, Takanori Yamazaki, Masayuki Shiota, Soichi Sano, Masako Tanaka, Mayuko Osada-Oka, Kenei Shimada, Katuyuki Miura, Minoru Yoshiyama, Hiroshi Iwao
ABSTRACT
Remote ischemic conditioning (RIC) by repeated treatment of transient limb ischemia is a clinically applicable method for protecting the heart against injury at the time of reperfusion. In this study, we investigated the effects of repeated RIC on cardiac dysfunction after myocardial infarction (MI). At 4weeks after MI, rats were separated into the untreated (UT) group or the RIC-treated group. RIC treatment was performed by 5cycles of 5min of bilateral hindlimb ischemia and 5min of reperfusion once a day for 4weeks. Despite comparable MI size, left ventricular (LV) ejection fraction (LVEF) was significantly improved in the RIC group compared with the UT group. Furthermore, the LVEF in the RIC group was improved, although not significantly, after treatment. RIC treatment also prevented the deterioration of LV diastolic function. MI-induced LV interstitial fibrosis in the boundary region and oxidant stress were significantly attenuated by RIC treatment. MicroRNA-29a (miR-29a), a key regulator of tissue fibrosis, was highly expressed in the exosomes and the marginal area of the RIC group. Even in the differentiated C2C12-derived exosomes, miR-29a expression was significantly increased under hypoxic condition. As well as miR-29a, insulin-like growth factor 1 receptor (IGF-1R) was highly expressed both in the exosomes and remote non-infarcted myocardium of the RIC group. IGF-1R expression was also increased in the C2C12-derived exosomes under hypoxic conditions. Repeated RIC reduces adverse LV remodeling and oxidative stress by MI. Exosome-mediated intercellular communication may contribute to the beneficial effect of RIC treatment.
Product Description
SAFC
Sodium deoxycholate
Sigma-Aldrich
Sodium fluoride, puriss., meets analytical specification of Ph. Eur., BP, USP, 98.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
Supelco
Sodium Fluoride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium fluoride, 99.99% trace metals basis
Sigma-Aldrich
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
Supelco
Fluoride ion solution for ISE, 0.1 M F-, for ion-selective electrodes
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Sodium fluoride, BioXtra, ≥99%
Sigma-Aldrich
Sodium fluoride, BioReagent, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Sodium fluoride 0.5 M solution
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, ≥97.0%
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%