Merck
  • Effect of lyophilization on liposomal encapsulation of salmon calcitonin.
Effect of lyophilization on liposomal encapsulation of salmon calcitonin.
Journal of liposome research (2014-04-09)
Janani Swaminathan, Carsten Ehrhardt
ABSTRACT
The intent of this work was to assess the impact of lyophilization on the encapsulation of salmon calcitonin (sCT) into liposomes. Four different liposomal formulations were investigated, i.e. DPPC:Chol:DSPE-PEG(2000) (75:20:5 and 65:30:5) and DPPC:Chol (80:20 and 66.7:33.3). Lipid films were prepared and hydrated with loading buffer containing sCT and different concentrations of the cryoprotectant, trehalose dihydrate. The liposomes were lyophilized, reconstituted and extruded to obtain small unilamellar vesicles. Non-encapsulated sCT was separated by gel filtration. Non-lyophilized formulations and liposomes lyophilized without the cryoprotectant were used as controls. Liposomes were analyzed for particle size, polydispersity index, zeta-potential and encapsulation efficiency. ³¹P-NMR (phosphorous nuclear magnetic resonance spectroscopy) was performed on selected formulations. Post-lyophilization, no significant change in particle sizes and zeta-potentials were noted, regardless of the presence or absence of the cryoprotectant. Encapsulation efficiencies, however, increased following lyophilization, in both PEGylated (lyophilization control batch) and non-PEGylated liposomes (cryoprotectant batches only). ³¹P-NMR revealed the presence of two distinct vesicle populations--liposomes and micelles--in PEGylated formulation. The presence of micelles might be responsible for the observed encapsulation enhancement of sCT in the PEGylated formulation. Lyophilization resulted in an increase in encapsulation efficiency of sCT in PEGylated liposomes, even in the absence of a cryoprotectant, due to presence of micellar vesicles.
Sigma-Aldrich
Potassium chloride, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Potassium chloride, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Potassium chloride solution, conductance standard C acc. to ISO 7888, 0.001 M KCl
Sigma-Aldrich
Sodium bicarbonate, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Supelco
Potassium chloride solution, conductance standard A acc. to ISO 7888, 0.1 M KCl
Supelco
Potassium chloride solution, conductance standard B acc. to ISO 7888, 0.01 M KCl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium phosphate monobasic-16O4, 99.9 atom % 16O
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Potassium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
SAFC
Sodium chloride solution, 5 M
Trehalose dihydrate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium bicarbonate, BioXtra, 99.5-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)