Merck
  • In vivo two-photon imaging of structural dynamics in the spinal dorsal horn in an inflammatory pain model.
In vivo two-photon imaging of structural dynamics in the spinal dorsal horn in an inflammatory pain model.
The European journal of neuroscience (2015-02-04)
Shinji Matsumura, Wataru Taniguchi, Kazuhiko Nishida, Terumasa Nakatsuka, Seiji Ito
ABSTRACT
Two-photon microscopy imaging has recently been applied to the brain to clarify functional and structural synaptic plasticity in adult neural circuits. Whereas the pain system in the spinal cord is phylogenetically primitive and easily exhibits behavioral changes such as hyperalgesia in response to inflammation, the structural dynamics of dendrites has not been analysed in the spinal cord mainly due to tissue movements associated with breathing and heart beats. Here we present experimental procedures to prepare the spinal cord sufficiently to follow morphological changes of neuronal processes in vivo by using two-photon microscopy and transgenic mice expressing fluorescent protein specific to the nervous system. Structural changes such as the formation of spine-like structures and swelling of dendrites were observed in the spinal dorsal horn within 30 min after the multiple-site injections of complete Freund's adjuvant (a chemical irritant) to a leg, and these changes continued for 5 h. Both AMPA and N-methyl-D-aspartate receptor antagonists, and gabapentin, a presynaptic Ca(2+) channel blocker, completely suppressed the inflammation-induced structural changes in the dendrites in the spinal dorsal horn. The present study first demonstrated by in vivo two-photon microscopy imaging that structural synaptic plasticity occurred in the spinal dorsal horn immediately after the injection of complete Freund's adjuvant and may be involved in inflammatory pain. Furthermore, acute inflammation-associated structural changes in the spinal dorsal horn were shown to be mediated by glutamate receptor activation.
Product Description
USP
Potassium gluconate, United States Pharmacopeia (USP) Reference Standard
Supelco
Potassium gluconate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Potassium gluconate, 97.0-103.0% dry basis, meets USP testing specifications
Sigma-Aldrich
DNQX, ≥98% (TLC)
Sigma-Aldrich
Potassium D-gluconate, ≥99% (HPLC)
Sigma-Aldrich
Anti-Post Synaptic Density Protein 95 Antibody, clone 6G6-1C9, clone 6G6-1C9, Chemicon®, from mouse
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Supelco
Sodium chloride, Pharmaceutical Secondary Standard; Certified Reference Material