Skip to Content
Merck

AB10181

Anti-Pumilio-2 (PUM2) Antibody

from rabbit, purified by affinity chromatography

Synonym(s):

pumilio homolog 2 (Drosophila), pumilio homolog 2, pumilio (Drosphila) homolog 2

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

rhesus macaque, mouse, human

species reactivity (predicted by homology)

chimpanzee (based on 100% sequence homology), rhesus monkey (based on 100% sequence homology), canine (based on 100% sequence homology)

technique(s)

immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... PUM2(23369)

General description

PUM2 is a member of the Puf family of RNA-binding proteins, which are believed to play a role in the regulation of posttranscriptional gene expression. Members of the Puf family of RNA binding proteins function as repressors of translation when bound to the 3-untranslated region of RNA transcripts.
~120 kDa observed.
Non specific band may be observed at 48 kDa

Immunogen

Epitope: N-terminus
KLH-conjugated linear peptide corresponding to the N-terminus of human Pumilio-2.

Application

Detect Pumilio-2 (PUM2) using this Anti-Pumilio-2 (PUM2) Antibody validated for use in IC, IH & WB.
Immunohistochemistry Analysis: A 1:400 dilution from a previous lot detected Pumilio-2 in human testis and teratoma tissue.

Immunocytochemistry Analysis: A 1:500 dilution from a previous lot detected Pumilio-2 in human and mouse embryonic stem cells.
Research Category
Stem Cell Research
Research Sub Category
RNA Metabolism & Binding Proteins

Biochem/physiol Actions

This antibody recognizes Pumilio-2 near the N-terminus.

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4, 150 mM NaCl) with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse embryonic stem cell lysate
Evaluated by Western Blot on mouse embryonic stem cell lysate.

Western Blot Analysis: 1 µg/mL of this antibody detected Pumilio-2 (PUM2) in 10 µg of mouse embryonic stem cell lysate.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service