Skip to Content
Merck

AB15564-200UL

Anti-proNT-3 Antibody

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

conjugate

unconjugated

Quality Level

antibody form

affinity purified immunoglobulin

clone

polyclonal

purified by

(ImmunoAffinity Purified)

species reactivity

rabbit, rat, mouse

technique(s)

immunohistochemistry: suitable
western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

FUNCTION: SwissProt: P20783 # Seems to promotes the survival of visceral and proprioceptive sensory neurons.
SIZE: 257 amino acids; 29355 Da
SUBCELLULAR LOCATION: Secreted.
TISSUE SPECIFICITY: Brain and peripheral tissues.
SIMILARITY: SwissProt: P20783 ## Belongs to the NGF-beta family.
Purified peptide from the pro domain of the human NT-3 protein (Accession number P20783).

Application

Western blot on recombinant mouse proNT-3
Immunohistochemistry on mouse brain sections.
Dilutions should be made using a carrier protein such as BSA (1-3%).

Optimal working dilutions must be determined by the end user.

Biochem/physiol Actions

Recognizes proNT-3 (pro Neurotrophin-3).

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% BSA, and 0.05% sodium azide as a preservative. Reconstitute with 200 μL of sterile deionized water. Centrifuge antibody preparation before use (10,000 xg for 5 min).

Preparation Note

Maintain lyophilized material at -20°C for up to 12 months after date of receipt. After reconstitution maintain at -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Other Notes

Included free of charge with the antibody is 40 μg of control antigen (lyophilized powder). The stock solution of the antigen can be made up using 100 μL of sterile deionized water. For negative control, preincubate 1 μg of peptide with 1 μg of antibody for one hour at room temperature. Optimal concentrations must be determined by the end user.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service