Skip to Content
Merck

ZRB2366

Anti-p-SMC1A (Ser966) Antibody, clone 1B11 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

SMC protein 1A, SMC-1-alpha, SMC-1A, Sb1.8, Structural maintenance of chromosomes protein 1A

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1B11, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 143.23 kDa
observed mol wt ~143 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
inhibition assay: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... SMC1A(8243)

General description

Structural maintenance of chromosomes protein 1A (UniProt: Q14683; also known as SMC protein 1A, SMC-1-alpha, SMC-1A, Sb1.8) is encoded by the SMC1A (also known as DXS423E, KIAA0178, SB1.8, SMC1, SMC1L1) gene (Gene ID: 8243) in human. SMC1 is a ATPase and subunit of the multiprotein cohesion complex, a protein involved in chromosome segregation and in double-strand DNA break (DSB) repair. The cohesion multiprotein complex is required for sister chromatid cohesion. This complex is composed partly of two structural maintenance of chromosomes (SMC) proteins, SMC3 and either SMC1B or the protein encoded by this gene. Most of the cohesion complexes dissociate from the chromosomes before mitosis, although those complexes at the kinetochore remain. Therefore, the encoded protein is thought to be an important part of functional kinetochores. Structurally, SMC1A consists of five distinct regions: two coiled-coil domains, a hinge domain, and N- and C-terminal domains, which include ATP and DNA binding motifs. It is ubiquitously expressed, with higher levels found in lymph nodes, appendix, and colon. SMC1A is predominantly localized in the nucleus. The protein also interacts with BRCA1 and ATM proteins indicating a role in DNA repair. ATM phosphorylates SMC1 at Ser957 and Ser966 both in vitro and in vivo. SMC1A at Ser966 is also phosphorylated by ATR kinases. Phosphorylated SMC1 is part of the complex that functions as a downstream effector in both the ATM/NBS1 branch and the ATR/MSH2 branch of S-phase checkpoint. Mutations in this gene result in Cornelia de Lange syndrome. It is reportedly involved in the G2/M transition of human glioma cells and G1/S transition of human lung adenocarcinoma cells. However, recent studies have also suggested its pivotal role in epithelial mesenchymal transition, tumorigenesis, metastasis, and chemo- and radio-resistance in cancer cells. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yadav, S., et al. (2025). Cells. 14(2);128; Liu, Y., et al. (2023). BMC Gastroenterol. 23; Article 268).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 966 from the C-terminal half of human SMC1A.

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cells treated with Etoposide.

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SMC1A phosphorylated at serine 966 in lysate from HeLa cells treated with Etoposide (20 g/mL; 3 h) but, not in untreated HeLa cell lysate.

Tested Applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected SMC1A phosphorylated at Ser966 in HeLa cells treated with Etoposide (20 g/mL for 3 hours, post serum starvation overnight) but, not in untreated HeLa cells.

Peptide Inhibition Assay: Target band detection in lysate from HeLa cells treated with Etoposide (20 g/mL; 3 h) was prevented by pre-blocking of a representative lot with the immunogen phospho-peptide, but not with the corresponding non-phospho peptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 1B11 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects SMC1A phosphorylated at serine 966.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service