Skip to Content
Merck

ZRB3213

Anti-QKI Antibody, clone 1J10 ZooMAb® Rabbit Monoclonal

newenhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Hqk, HqkI, KH domain-containing RNA-binding protein QKI, Protein quaking

Sign In to View Organizational & Contract Pricing

Select a Size


About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J10, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 37.67 kDa
observed mol wt ~38 kDa

purified by

using Protein A

species reactivity

rat, human, mouse

species reactivity (predicted by homology)

zebrafish

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

Internal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... QKI(9444)

General description

KH domain-containing RNA-binding protein QKI (UniProt: Q96PU8; also known as Protein quaking, Hqk, HqkI) is encoded by the QKI (also known as HKQ) gene (Gene ID: 9444) in human. The gene QKI (quaking) is mapped to human chromosome 6q26. It is a RNA reader protein, which recognizes and binds specific RNAs, and belongs to signal transduction and activation of RNA (STAR) protein family. QKI acts by regulating pre-mRNA splicing, mRNA export, mRNA stability and protein translation, and is itself, regulated by alternative splicing. QKI contains a KH RNA-binding domain flanked by two QUA domains, which facilitate dimerization and RNA interaction, crucial for its regulatory functions in alternative splicing, mRNA stability, and translation. Six isoforms of QKI have been described that are produced by alternative splicing but, exists predominantly as three major isoforms; QKI-5, QKI-6, and QKI-7, each differing in their C-terminal regions and subcellular localization, primarily within the nucleus and cytoplasm. QKI is expressed in the frontal cortex of brain, but is shown to be down-regulated in the brain of schizophrenic patients. Aberrant expression of QKI has been implicated in various diseases, including multiple sclerosis, schizophrenia, and certain cancers, where it may act as either a tumor suppressor or an oncogene depending on the context. Mouse homolog of QKI participates in neural development and myelination. Deficiencies in QKI are linked to severe demyelination. It also controls the production of circRNAs during epithelial-mesenchymal transition. In lung cancer cells, it suppresses proliferation by regulating alternative splicing of NUMB (protein S171) gene. QKI has also been demonstrated as a transcriptional co-activator beyond its canonical function as a RNA-binding protein. This ZooMAb® recombinant monoclonal antibody, generated by our proprietary technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Neumann, DP., et al. (2022). WIREs RNA. 13(6); 1724; Zhou, X., et al. (2021). elife. Dev Bio. Neuroscience. 10; 60467).
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the internal region of human QKI.

Biochem/physiol Actions

Clone 1J10 is a ZooMAb® recombinant rabbit monoclonal antibody that specifically detects QKI. It targets an epitope within 19 amino acids from the internal region. This antibody is predicted to recognize QKI-6, QKI-7, and QKI-7B isoforms.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Preparation Note

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

We are committed to bringing you greener alternative products, which adhere to one or more of the 12 Principles of Green Chemistry. This antibody is preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed, and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service