MilliporeSigma
HomeWebinarsBoost Efficiency and Sustainability of your Listeria Testing
Boost Efficiency and Sustainability of your Listeria Testing
Webinar
Join us for our webinar on the ReadyStream® system, a revolutionary solution for culture media preparation and dispensing. After providing a general overview on the working principle and features of the ReadyStream® system, this session will focus on our newly launched, and first-of-its kind 5x concentrated ReadyStream® Half FRASER medium, used for the primary selective enrichment of Listeria monocytogenes and Listeria spp. according to ISO 11290.
The ReadyStream® system allows for the delivery of large volumes of Buffered Peptone Water (BPW), Half FRASER, and Tryptic Soy Broth (TSB) with minimal preparation time, at a desired volume, temperature, and concentration for pre-enrichment of samples.
Discover how this innovative system works and the numerous benefits it offers in terms of efficiency, convenience, and sustainability through reduced energy, water consumption, as well as significantly less plastic waste.
In this webinar, you will learn:
  • How the ReadyStream® system simplifies culture media preparation and dispensing.
  • The benefits of using this innovative system over traditional methods
  • Overview of the new ReadyStream® Half FRASER medium for Listeria detection
The webinar is also available in six additional languages: French, German, Italian, Spanish, Portuguese, and Chinese.
Speaker
Markus Prengel
Markus Prengel
Merck KGaA, Darmstadt, Germany
Global Product Manager, Innovation Food & Beverage
Markus Prengel holds a master’s degree in business chemistry and is based in Darmstadt, Germany. He first spent two years in the Quality Department of the Life Science division of Merck KGaA, Darmstadt, Germany, and has served as a Global Product Manager for Pharma and Food & Beverage Microbiology QC Products for the past four years.