Skip to Content
MilliporeSigma

BDPH9BCCD439

Bide Pharmatech

3-(4-Aminophenyl)-1,1,3-trimethyl-2,3-dihydro-1H-inden-5-amine

Synonym(s):

3-(4-aminophenyl)-1,1,3-trimethyl-5-indanamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C18H22N2
CAS Number:
Molecular Weight:
266.39
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C18H22N2/c1-17(2)11-18(3,12-4-6-13(19)7-5-12)16-10-14(20)8-9-15(16)17/h4-10H,11,19-20H2,1-3H3

Inchi Key

GDGWSSXWLLHGGV-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%56

Country of Origin

CN

Physical Form

Solid

Storage temp.

Keep in dark place, sealed in dry, 2-8°C



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library