Skip to Content
MilliporeSigma

BDPH9BCCFCD5

Bide Pharmatech

AChE/BChE/MAO-B-IN-1

Synonym(s):

(1-benzylpiperidin-4-yl)methyl hydrogen phenylcarbonimidate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C20H24O2N2
CAS Number:
Molecular Weight:
324.42
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C20H24N2O2/c23-20(21-19-9-5-2-6-10-19)24-16-18-11-13-22(14-12-18)15-17-7-3-1-4-8-17/h1-10,18H,11-16H2,(H,21,23)

Inchi Key

IKTRKOHDYGDICC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

Powder or crystals

Storage temp.

Keep in dark place, sealed in dry, 2-8°C



Still not finding the right product?

Explore all of our products under AChE/BChE/MAO-B-IN-1




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library