Skip to Content
MilliporeSigma

BDPH9BCD19F2

Bide Pharmatech

2-Iminothiolane hydrochloride

Synonym(s):

dihydro-2(3H)-thiophenimine hydrochloride

Select a Size

Change View

About This Item

Linear Formula:
C4H8N1Cl1S1
CAS Number:
Molecular Weight:
137.63
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C4H7NS.ClH/c5-4-2-1-3-6-4;/h2H,1,3,5H2;1H

Inchi Key

HIQWKSAWCPDJSL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Iminothiolane hydrochloride


hstatements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library