Skip to Content
MilliporeSigma

BDPH9BCD2D09

Bide Pharmatech

5-Hydroxy-2-methylisonicotinic acid

Synonym(s):

5-hydroxy-2-methylisonicotinic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H7O3N1
CAS Number:
Molecular Weight:
153.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H7NO3/c1-4-2-5(7(10)11)6(9)3-8-4/h2-3,9H,1H3,(H,10,11)

Inchi Key

PZFCGNPCQFHNCX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library