Skip to Content
MilliporeSigma

BDPH9BCD4BED

Bide Pharmatech

1-(4-Bromophenyl)butane-1,3-dione

Synonym(s):

(3Z)-4-(4-bromophenyl)-4-hydroxy-3-buten-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H9O2Br1
CAS Number:
Molecular Weight:
241.08
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H9BrO2/c1-7(12)6-10(13)8-2-4-9(11)5-3-8/h2-5H,6H2,1H3

Inchi Key

GIKXMINIUUFRFD-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Country of Origin

CN

Physical Form

White to Yellow Solid

Storage temp.

Inert atmosphere, 2-8°C



Still not finding the right product?

Explore all of our products under 1-(4-Bromophenyl)butane-1,3-dione


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library