Skip to Content
MilliporeSigma

BDPH9BCD4E08

Bide Pharmatech

[1,2,4]Triazolo[4,3-a]pyridine

Synonym(s):

[1,2,4]triazolo[4,3-a]pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5N3
CAS Number:
Molecular Weight:
119.13
MDL number:
Product BDPH9BCD4E08 is not currently sold in your country.


Inchi Code

1S/C6H5N3/c1-2-4-9-5-7-8-6(9)3-1/h1-5H

Inchi Key

AYSYSOQSKKDJJY-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under [1,2,4]Triazolo[4,3-a]pyridine


signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library