Skip to Content
MilliporeSigma

BDPH9BCD7067

Bide Pharmatech

tert-Butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate

Synonym(s):

tert-butyl (2-(2-(2-iodoethoxy)ethoxy)ethyl)carbamate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H22O4N1I1
CAS Number:
Molecular Weight:
359.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C11H22INO4/c1-11(2,3)17-10(14)13-5-7-16-9-8-15-6-4-12/h4-9H2,1-3H3,(H,13,14)

Inchi Key

LRJHSBOQFJGFFX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid or liquid



Still not finding the right product?


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library