Skip to Content
MilliporeSigma

BDPH9BCD718E

Bide Pharmatech

[1,1'-Biphenyl]-3,3',4,4'-tetraamine

Synonym(s):

[1,1'-biphenyl]-3,3',4,4'-tetramine

Select a Size

Change View

About This Item

Linear Formula:
C12H14N4
CAS Number:
Molecular Weight:
214.27
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C12H14N4/c13-9-3-1-7(5-11(9)15)8-2-4-10(14)12(16)6-8/h1-6H,13-16H2

Inchi Key

HSTOKWSFWGCZMH-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

98%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Pale yellow to Greyish yellow to Brown Solid

Melting Point

175-177C



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library