Skip to Content
MilliporeSigma

BDPH9BCDBEBB

Bide Pharmatech

(2-Bromo-6-chloropyridin-3-yl)methanol

Synonym(s):

(2-bromo-6-chloropyridin-3-yl)methanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H5O1N1Cl1Br1
CAS Number:
Molecular Weight:
222.47
MDL number:
Product BDPH9BCDBEBB is not currently sold in your country.


Inchi Code

1S/C6H5BrClNO/c7-6-4(3-10)1-2-5(8)9-6/h1-2,10H,3H2

Inchi Key

CUVPULOYOCRYDC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library