Skip to Content
MilliporeSigma

BDPH9C0212DB

Bide Pharmatech

(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-6-(((4-Methoxyphenyl)diphenylmethyl)amino)hexanoic acid

Synonym(s):

(2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-6-{[(4-methoxyphenyl)(diphenyl)methyl]amino}hexanoic acid

Select a Size

Change View

About This Item

Linear Formula:
C41H40O5N2
CAS Number:
Molecular Weight:
640.78
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C41H40N2O5/c1-47-32-25-23-31(24-26-32)41(29-14-4-2-5-15-29,30-16-6-3-7-17-30)42-27-13-12-22-38(39(44)45)43-40(46)48-28-37-35-20-10-8-18-33(35)34-19-9-11-21-36(34)37/h2-11,14-21,23-26,37-38,42H,12-13,22,27-28H2,1H3,(H,43,46)(H,44,45)/t38-/m0/s1

Inchi Key

CTYHQVFFQRDJSN-LHEWISCISA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Solid





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library