Skip to Content
MilliporeSigma

PG9H9BC7B20F

PrecisePEG

Amido Mal-PEG4-NHS Ester

Synonym(s):

3-(2,5-dioxo-2,5-dihydro-1H-pyrrol-1-yl)-N-{15-[(2,5-dioxo-1-pyrrolidinyl)oxy]-15-oxo-3,6,9,12-tetraoxapentadec-1-yl}propanamide

Select a Size

Change View

About This Item

Linear Formula:
C22H31O11N3
CAS Number:
Molecular Weight:
513.5
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C22H31N3O11/c26-17(5-8-24-18(27)1-2-19(24)28)23-7-10-33-12-14-35-16-15-34-13-11-32-9-6-22(31)36-25-20(29)3-4-21(25)30/h1-2H,3-16H2,(H,23,26)

Inchi Key

XSRYGQJQNPJNKZ-UHFFFAOYSA-N

Shipping Temperature

ambient

Purity

90%

Storage temp.

-20

Country of Origin

CN

Physical Form

Oil



hstatements

Precautionary Statements



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library