Skip to Content
Products
Cart
0
CA
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Buffer pH 10.0 Blue
Products
Product Category
Physical & Chemical Property Standards
reference materials
buffers
Brand
Supelco
Quality Segment
Application
Special Grade
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
BX16331
Buffer pH 10.0 Blue,
certified reference material
Expand
View Pricing
BX16411
Buffer pH 10.0 Blue,
certified reference material
Expand
View Pricing
An unknown error has occured.