Skip to Content
Products
Cart
0
CA
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Buffer pH 1.68
Products
Product Category
Physical & Chemical Property Standards
buffers
reference materials
Brand
Supelco
Quality Segment
Application
Special Grade
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
EM01822
Buffer pH 1.68,
certified reference material
Expand
View Pricing
An unknown error has occured.