Skip to Content
Products
Cart
0
CA
EN
Products
Products
Applications
Services
Resources
Support
Login / Register
Order Lookup
Quick Order
Cart
0
Back
Sodium hydroxide-
16
O solution
Linear Formula:
Na
16
OH
Molecular Weight:
39.99
Products
Product Category
bases
stable isotopes
Brand
Sigma-Aldrich
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
609765
Sodium hydroxide-
16
O solution,
20 wt. % in H
2
16
O, 99.9 atom %
16
O
Expand
View Pricing
An unknown error has occured.