MilliporeSigma
  • Comparison of the results obtained by ELISA and surface plasmon resonance for the determination of antibody affinity.
Comparison of the results obtained by ELISA and surface plasmon resonance for the determination of antibody affinity.
Journal of Immunological Methods (2010)
Heinrich L, et al.