MilliporeSigma
  • Rett syndrome mutations abolish the interaction of MeCP2 with the NCoR/SMRT co-repressor.
Rett syndrome mutations abolish the interaction of MeCP2 with the NCoR/SMRT co-repressor.
Nature neuroscience (2013-06-19)
Matthew J Lyst, Robert Ekiert, Daniel H Ebert, Cara Merusi, Jakub Nowak, Jim Selfridge, Jacky Guy, Nathaniel R Kastan, Nathaniel D Robinson, Flavia de Lima Alves, Juri Rappsilber, Michael E Greenberg, Adrian Bird
ABSTRACT
Rett syndrome (RTT) is a severe neurological disorder that is caused by mutations in the MECP2 gene. Many missense mutations causing RTT are clustered in the DNA-binding domain of MeCP2, suggesting that association with chromatin is critical for its function. We identified a second mutational cluster in a previously uncharacterized region of MeCP2. We found that RTT mutations in this region abolished the interaction between MeCP2 and the NCoR/SMRT co-repressor complexes. Mice bearing a common missense RTT mutation in this domain exhibited severe RTT-like phenotypes. Our data are compatible with the hypothesis that brain dysfunction in RTT is caused by a loss of the MeCP2 'bridge' between the NCoR/SMRT co-repressors and chromatin.
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, clone M2, purified immunoglobulin (Purified IgG1 subclass), buffered aqueous solution (10 mM sodium phosphate, 150 mM NaCl, pH 7.4, containing 0.02% sodium azide)
Sigma-Aldrich
Anti-NeuN Antibody, clone A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
Anti-MeCP2 antibody, Mouse monoclonal, clone Men-8, purified from hybridoma cell culture
Sigma-Aldrich
Anti-MeCP2 antibody, Mouse monoclonal, clone Mec-168, purified from hybridoma cell culture