MilliporeSigma
  • A membrane-permeable peptide containing the last 21 residues of the G alpha(s) carboxyl terminus inhibits G(s)-coupled receptor signaling in intact cells: correlations between peptide structure and biological activity.
A membrane-permeable peptide containing the last 21 residues of the G alpha(s) carboxyl terminus inhibits G(s)-coupled receptor signaling in intact cells: correlations between peptide structure and biological activity.
Molecular pharmacology (2005-12-08)
Anna Maria D'Ursi, Laura Giusti, Stefania Albrizio, Francesca Porchia, Cinzia Esposito, Gabriella Caliendo, Claudia Gargini, Ettore Novellino, Antonio Lucacchini, Paolo Rovero, Maria Rosa Mazzoni
ABSTRACT
Cell-penetrating peptides are able to transport covalently attached cargoes such as peptide or polypeptide fragments of endogenous proteins across cell membranes. Taking advantage of the cell-penetrating properties of the 16-residue fragment penetratin, we synthesized a chimeric peptide that possesses an N-terminal sequence with membrane-penetrating activity and a C-terminal sequence corresponding to the last 21 residues of G alpha(s). This G alpha(s) peptide was an effective inhibitor of 5'-N-ethylcarboxamidoadenosine (NECA) and isoproterenol-stimulated production of cAMP in rat PC12 and human microvascular endothelial (HMEC-1) cells, whereas the carrier peptide had no effect. The maximal efficacy of NECA was substantially reduced when PC12 cells were treated with the chimeric peptide, suggesting that it competes with G alpha(s) for interaction with receptors. The peptide inhibited neither G(q)- nor G(i)-coupled receptor signaling. The use of a carboxy-fluorescein derivative of the peptide proved its ability to cross the plasma membrane of live cells. NMR analysis of the chimeric peptide structure in a membrane-mimicking environment showed that the G alpha(s) fragment assumed an amphipathic alpha-helical conformation tailored to make contact with key residues on the intracellular side of the receptor. The N-terminal penetratin portion of the molecule also showed an alpha-helical structure, but hydrophobic and hydrophilic residues formed clustered surfaces at the N terminus and center of the fragment, suggesting their involvement in the mechanism of penetratin internalization by endocytosis. Our biological data supported by NMR analysis indicate that the membrane-permeable G alpha(s) peptide is a valuable, nontoxic research tool to modulate G(s)-coupled receptor signal transduction in cell culture models.
Sigma-Aldrich
cAMP Enzyme Immunoassay Kit, Direct
Pricing and availability is not currently available.