MilliporeSigma
  • Intraspinal administration of interleukin-7 promotes neuronal apoptosis and limits functional recovery through JAK/STAT5 pathway following spinal cord injury.
Intraspinal administration of interleukin-7 promotes neuronal apoptosis and limits functional recovery through JAK/STAT5 pathway following spinal cord injury.
Biochemical and biophysical research communications (2019-05-10)
Xiaochen Yuan, Qingbin Wu, Peng Wang, Yingli Jing, Haijiang Yao, Yinshan Tang, Ruiqin Han, Weili He, Zhigang Li, Honggang Zhang, Ruijuan Xiu
ABSTRACT
It has been previously reported that the blockade of interleukin-7 receptor (IL-7R) promotes functional recovery following spinal cord injury (SCI), however, the direct function and molecular mechanism of IL-7 involved in this pathogenic process are unclear. Here, we report that, contrary to IL-7R blockade, the intraspinal administration of IL-7 limits functional recovery following SCI. In addition, IL-7 treatment promotes neuronal apoptosis in spinal cord lesions, which may be attributed to exacerbated focal inflammatory response, as shown by increased accumulation of activated microglia/macrophage and production of proinflammatory mediators. Moreover, IL-7 treatment activates JAK/STAT5 pathway following SCI. At last, more importantly, the pharmacological inhibition of STAT5 abrogates the effects of IL-7 treatment on functional recovery, neuronal apoptosis and focal inflammatory response, suggesting that the effects of IL-7 treatment following SCI are dependent on activating the JAK/STAT5 pathway. Overall, this study reveals the JAK/STAT5 pathway-dependent detrimental role of IL-7 following SCI, and also implies that targeting the IL-7/JAK/STAT5 axis may represent a potential therapeutic approach for SCI treatment.