MilliporeSigma
  • Additive manufacturing of photo-crosslinked gelatin scaffolds for adipose tissue engineering.
Additive manufacturing of photo-crosslinked gelatin scaffolds for adipose tissue engineering.
Acta biomaterialia (2019-05-29)
Liesbeth Tytgat, Lana Van Damme, Jasper Van Hoorick, Heidi Declercq, Hugo Thienpont, Heidi Ottevaere, Phillip Blondeel, Peter Dubruel, Sandra Van Vlierberghe
ABSTRACT
There exists a clear clinical need for adipose tissue reconstruction strategies to repair soft tissue defects which outperform the currently available approaches. In this respect, additive manufacturing has shown to be a promising alternative for the development of larger constructs able to support adipose tissue engineering. In the present work, a thiol-ene photo-click crosslinkable gelatin hydrogel was developed which allowed extrusion-based additive manufacturing into porous scaffolds. To this end, norbornene-functionalized gelatin (Gel-NB) was combined with thiolated gelatin (Gel-SH). The application of a macromolecular gelatin-based thiolated crosslinker holds several advantages over conventional crosslinkers including cell-interactivity, less chance at phase separation between scaffold material and crosslinker and the formation of a more homogeneous network. Throughout the paper, these photo-click scaffolds were benchmarked to the conventional methacrylamide-modified gelatin (Gel-MA). The results indicated that stable scaffolds could be realized which were further characterized physico-chemically by performing swelling, mechanical and in vitro biodegradability assays. Furthermore, the seeded adipose tissue-derived stem cells (ASCs) remained viable (>90%) up to 14 days and were able to proliferate. In addition, the cells could be differentiated into the adipogenic lineage on the photo-click crosslinked scaffolds, thereby performing better than the cells supported by the frequently reported Gel-MA scaffolds. As a result, the developed photo-click crosslinked scaffolds can be considered a promising candidate towards adipose tissue engineering and a valuable alternative for the omnipresent Gel-MA. STATEMENT OF SIGNIFICANCE: The field of adipose tissue engineering has emerged as a promising strategy to repair soft tissue defects. Herein, Gel-NB/Gel-SH gelatin-based hydrogel scaffolds were produced using extrusion-based additive manufacturing. Using a cell-interactive, thiolated gelatin crosslinker, a homogeneous network was formed and the risk of phase separation between norbornene-modified gelatin and macromolecular crosslinkers was reduced. UV-induced crosslinking of these materials is based on step growth polymerization which requires less free radicals to enable polymerization. Our results demonstrated the potential of the developed scaffolds, due to their favourable physico-chemical characteristics as well as their adipogenic differentiation potential when benchmarked to Gel-MA scaffolds. Hence, the hydrogels could be of great interest towards future development of adipose tissue constructs and tissue engineering in general.