MilliporeSigma
  • Expression, Localization, and Regulation of the Sodium Bicarbonate Cotransporter NBCe1 in the Thyroid.
Expression, Localization, and Regulation of the Sodium Bicarbonate Cotransporter NBCe1 in the Thyroid.
Thyroid : official journal of the American Thyroid Association (2018-12-12)
Myrna Virreira, Ling Jin, Sami Djerbib, Xavier De Deken, Françoise Miot, Claude Massart, Michal Svoboda, Jacqueline Van Sande, Renaud Beauwens, Jacques-Emile Dumont, Alain Boom
ABSTRACT
The intrafollicular space of thyroid follicles is the storage compartment for thyroid hormones. Its pH has been established at around 7.6 at least after thyrotropin (TSH) stimulation. This alkaline intrafollicular pH is thought to be critical for iodide coupling to thyroglobulin and internalization of iodinated thyroglobulin. At least in mice, this alkalinization requires the expression of pendrin (Slc26a4) within the apical membrane, and a lack of pendrin results in acidic follicular lumen pH. Yet, the mechanism importing HCO3- into the cytoplasm is unknown. This study investigated whether the rather ubiquitous sodium bicarbonate cotransporter NBCe1 (SLC4A4) might play this role. It also examined which variant was expressed and where it was localized in both rat and human thyroid tissue. Lastly, the dependence of its expression on TSH was studied. Reverse transcription polymerase chain reaction, immunofluorescence, and Western blotting were used to test whether TSH stimulated NBCe1 protein expression in vivo. Subcellular localization of NBCe1 was performed using immunofluorescence in both rat and human thyroid. Cultured thyroid cells were also used to attempt to define how TSH affects NBCe1 expression. Only transcripts of the NBCe1-B variant were detected in both rat and human thyroid. Of interest, NBCe1-C was not detected in human tissues, not even in the brain. On immunofluorescence microscopy, the immunostaining of NBCe1 mainly appeared in the basolateral membrane upon stimulation with TSH. This TSH induction of basolateral membrane expression of NBCe1 protein was confirmed in vivo in rat thyroid and in vitro on human thyroid slices. This study demonstrates the expression of the sodium bicarbonate cotransporter NBCe1-B in rat and human thyroid. Additionally, the data suggest that TSH blocks the degradation of NBCe1 protein by trafficking it to the basolateral membrane. Hence, TSH increases NBCe1 half-life without increasing its synthesis.