MilliporeSigma
  • A Label-Free Fluorescent Assay for the Rapid and Sensitive Detection of Adenosine Deaminase Activity and Inhibition.
A Label-Free Fluorescent Assay for the Rapid and Sensitive Detection of Adenosine Deaminase Activity and Inhibition.
Sensors (Basel, Switzerland) (2018-08-01)
Xinxing Tang, Kefeng Wu, Han Zhao, Mingjian Chen, Changbei Ma
ABSTRACT
Adenosine deaminase (ADA), able to catalyze the irreversible deamination of adenosine into inosine, can be found in almost all tissues and plays an important role in several diseases. In this work, we developed a label-free fluorescence method for the detection of adenosine deaminase activity and inhibition. In the presence of ADA, ATP has been shown to be hydrolyzed. The ATP aptamer was shown to form a G-quadruplex/thioflavin T (ThT) complex with ThT and exhibited an obvious fluorescence signal. However, the ATP aptamer could bind with ATP and exhibited a low fluorescence signal because of the absence of ADA. This assay showed high sensitivity to ADA with a detection limit of 1 U/L based on an SNR of 3 and got a good linear relationship within the range of 1⁻100 U/L with R² = 0.9909. The LOD is lower than ADA cutoff value (4 U/L) in the clinical requirement and more sensitive than most of the reported methods. This technique exhibited high selectivity for ADA against hoGG I, UDG, RNase H and λexo. Moreover, this strategy was successfully applied for assaying the inhibition of ADA using erythro-9-(2-hydroxy-3-nonyl) adenine (EHNA) and, as such, demonstrated great potential for the future use in the diagnosis of ADA-relevant diseases, particularly in advanced drug development.