MilliporeSigma
  • Integrated use of LC/MS/MS and LC/Q-TOF/MS targeted metabolomics with automated label-free microscopy for quantification of purine metabolites in cultured mammalian cells.
Integrated use of LC/MS/MS and LC/Q-TOF/MS targeted metabolomics with automated label-free microscopy for quantification of purine metabolites in cultured mammalian cells.
Purinergic signalling (2019-01-04)
S Eric Nybo, Jennifer T Lamberts
ABSTRACT
Purine metabolites have been implicated as clinically relevant biomarkers of worsening or improving Parkinson's disease (PD) progression. However, the identification of purine molecules as biomarkers in PD has largely been determined using non-targeted metabolomics analysis. The primary goal of this study was to develop an economical targeted metabolomics approach for the routine detection of purine molecules in biological samples. Specifically, this project utilized LC/MS/MS and LC/QTOF/MS to accurately quantify levels of six purine molecules in samples from cultured N2a murine neuroblastoma cells. The targeted metabolomics workflow was integrated with automated label-free digital microscopy, which enabled normalization of purine concentration per unit cell in the absence of fluorescent dyes. The established method offered significantly enhanced selectivity compared to previously published procedures. In addition, this study demonstrates that a simple, quantitative targeted metabolomics approach can be developed to identify and quantify purine metabolites in biological samples. We envision that this method could be broadly applicable to quantification of purine metabolites from other complex biological samples, such as cerebrospinal fluid or blood.