MilliporeSigma
  • Oblique-plane single-molecule localization microscopy for tissues and small intact animals.
Oblique-plane single-molecule localization microscopy for tissues and small intact animals.
Nature methods (2019-08-21)
Jeongmin Kim, Michal Wojcik, Yuan Wang, Seonah Moon, Emilia A Zin, Nadia Marnani, Zachary L Newman, John G Flannery, Ke Xu, Xiang Zhang
ABSTRACT
Single-molecule localization microscopy (SMLM), while well established for cultured cells, is not yet fully compatible with tissue-scale samples. We introduce single-molecule oblique-plane microscopy (obSTORM), which by directly imaging oblique sections of samples with oblique light-sheet illumination offers a deep and volumetric SMLM platform that is convenient for standard tissue samples and small intact animals. We demonstrate super-resolution imaging at depths of up to 66 µm for cells, Caenorhabditis elegans gonads, Drosophila melanogaster larval brain, mouse retina and brain sections, and whole stickleback fish.
Sigma-Aldrich
5-Bromo-2′-deoxyuridine, ≥99% (HPLC)
Sigma-Aldrich
Monoclonal Anti-α-Tubulin antibody produced in mouse, clone DM1A, ascites fluid