MilliporeSigma
  • Mass Spectrometric Detection and Characterization of Metabolites of Gemini Surfactants Used as Gene Delivery Vectors.
Mass Spectrometric Detection and Characterization of Metabolites of Gemini Surfactants Used as Gene Delivery Vectors.
Journal of the American Society for Mass Spectrometry (2020-01-11)
Wei Jin, Randy Purves, Ed Krol, Ildiko Badea, Anas El-Aneed
ABSTRACT
Gemini surfactants are a class of lipid molecules that have been successfully used in vitro and in vivo as nonviral gene delivery vectors. However, the biological fate of gemini surfactants has not been well investigated. In particular, the metabolism of gemini surfactants after they enter cells as gene delivery vehicles is unknown. In this work, we used a high-resolution quadrupole-Orbitrap mass spectrometry (Q-Exactive) instrument to detect the metabolites of three model gemini surfactants, namely, (a) unsubstituted (16-3-16), (b) with pyridinium head groups (16(Py)-S-2-S-16(Py)), and (c) substituted with a glycyl-lysine di-peptide (16-7N(GK)-16). The metabolites were characterized, and structures were proposed, based on accurate masses and characteristic product ions. The metabolism of the three gemini surfactants was very different as 16-3-16 was not metabolized in PAM 212 cells, whereas 16(Py)-S-2-S-16(Py) was metabolized primarily via phase I reactions, including oxidation and dealkylation, producing metabolites that could be linked to its observed high toxicity. The third gemini surfactant 16-7N(GK)-16 was metabolized mainly via phase II reactions, including methylation, acetylation, glucose conjugation, palmityl conjugation, and stearyl conjugation. The metabolism of gemini surfactants provides insight for future directions in the design and development of more effective gemini surfactants with lower toxicity. The reported approach can also be applied to study the metabolism of other structurally related gemini surfactants.
Product Description
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Avanti
18:1 (Δ9-Cis) PE (DOPE), Avanti Research - A Croda Brand