MilliporeSigma
  • Photocatalytic degradation of terephthalic acid using titania and zinc oxide photocatalysts: Comparative study
Photocatalytic degradation of terephthalic acid using titania and zinc oxide photocatalysts: Comparative study
Desalination (2010)
Shafaei A, et al.