MilliporeSigma
  • The peak-to-end of the T wave in the limb ECG leads reflects total spatial rather than transmural dispersion of ventricular repolarization in an anthopleurin-A model of prolonged QT interval.
The peak-to-end of the T wave in the limb ECG leads reflects total spatial rather than transmural dispersion of ventricular repolarization in an anthopleurin-A model of prolonged QT interval.
Heart rhythm (2011-11-30)
Daisuke Izumi, Masaomi Chinushi, Kenichi Iijima, Hiroshi Furushima, Yukio Hosaka, Kanae Hasegawa, Yoshifusa Aizawa
ABSTRACT
Previous studies have showed that the interval between the peak and the end of the T wave (Tp-e) is a marker of transmural dispersion of ventricular repolarization. We studied the relationship between (a) the Tp-e on local pseudo transmural electrograms (pseudo transmural Tp-e) or limb leads of body surface electrocardiogram (surface Tp-e) and (b) the intracardiac left ventricular (LV) repolarization during a drug-induced QT-interval prolongation. Using open-chested canine intact hearts treated by anthopleurin-A, transmural LV electrograms were recorded via needle electrodes placed in the basoanterior, midanterior, apicoanterior, basolateral, midlateral, and apicolateral LV wall. Recovery time (RT) was calculated as an index of local repolarization at each transmural unipolar electrode. This model showed slower heart rate-dependent heterogeneous distribution of ventricular repolarization both along the basal to apical axis and along the transmural axis. RT was longer at the LV apex than at the base and longer in the lateral than in the anterior wall during the slower heart rate. A high correlation was found between surface Tp-e and total LV dispersion. In contrast, pseudo transmural Tp-e correlated with transmural RT dispersion. The shortest RT in the heart roughly corresponded to the peak, as did the longest RT with the end of the T wave on the surface electrocardiogram. During drug-induced QT-interval prolongation with a large apicobasal and anterolateral dispersion of ventricular repolarization, the Tp-e in the limb leads expresses spatial (total) distribution of repolarization in the whole left ventricle.