MilliporeSigma
  • Unsaturated Fatty Acid-Induced Conformational Transitions and Aggregation of the Repeat Domain of Tau.
Unsaturated Fatty Acid-Induced Conformational Transitions and Aggregation of the Repeat Domain of Tau.
Molecules (Basel, Switzerland) (2020-06-18)
Carlo Giorgio Barracchia, Roberto Tira, Francesca Parolini, Francesca Munari, Luigi Bubacco, Georgios A Spyroulias, Mariapina D'Onofrio, Michael Assfalg
ABSTRACT
The intrinsically disordered, amyloidogenic protein Tau associates with diverse classes of molecules, including proteins, nucleic acids, and lipids. Mounting evidence suggests that fatty acid molecules could play a role in the dysfunction of this protein, however, their interaction with Tau remains poorly characterized. In a bid to elucidate the association of Tau with unsaturated fatty acids at the sub-molecular level, we carried out a variety of solution NMR experiments in combination with circular dichroism and fluorescence measurements. Our study shows that Tau4RD, the highly basic four-repeat domain of Tau, associates strongly with arachidonic and oleic acid assemblies in a high lipid/protein ratio, perturbing their supramolecular states and itself undergoing time-dependent structural adaptation. The structural signatures of Tau4RD/fatty acid aggregates appear similar for arachidonic acid and oleic acid, however, they are distinct from those of another prototypical intrinsically disordered protein, α-synuclein, when bound to these lipids, revealing protein-specific conformational adaptations. Both fatty acid molecules are found to invariably promote the self-aggregation of Tau4RD and of α-synuclein. This study describes the reciprocal influence that Tau4RD and fatty acids exert on their conformational states, contributing to our understanding of fundamental aspects of Tau/lipid co-assembly.
Sigma-Aldrich
Thioflavin T, used as stain for amyloid
Sigma-Aldrich
Heparin sodium salt from porcine intestinal mucosa, Grade I-A, ≥180 USP units/mg
Sigma-Aldrich
Sodium oleate, ≥99%
Sigma-Aldrich
Arachidonic acid sodium salt from Mortierella alpina, ≥98.5% (GC)