MilliporeSigma
Theileria apicoplast as a target for chemotherapy.
Antimicrobial agents and chemotherapy (2008-12-17)
Regina Lizundia, Dirk Werling, Gordon Langsley, Stuart A Ralph
ABSTRACT
Theileria parasites cause severe bovine disease and death in a large part of the world. These apicomplexan parasites possess a relic plastid (apicoplast), whose metabolic pathways include several promising drug targets. Putative inhibitors of these targets were screened, and we identified antiproliferative compounds that merit further characterization.
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)