MilliporeSigma
  • The Caulerpa pigment caulerpin inhibits HIF-1 activation and mitochondrial respiration.
The Caulerpa pigment caulerpin inhibits HIF-1 activation and mitochondrial respiration.
Journal of natural products (2009-11-20)
Yang Liu, J Brian Morgan, Veena Coothankandaswamy, Rui Liu, Mika B Jekabsons, Fakhri Mahdi, Dale G Nagle, Yu-Dong Zhou
ABSTRACT
The transcription factor hypoxia-inducible factor-1 (HIF-1) represents an important molecular target for anticancer drug discovery. In a T47D cell-based reporter assay, the Caulerpa spp. algal pigment caulerpin (1) inhibited hypoxia-induced as well as 1,10-phenanthroline-induced HIF-1 activation. The angiogenic factor vascular endothelial growth factor (VEGF) is regulated by HIF-1. Caulerpin (10 microM) suppressed hypoxic induction of secreted VEGF protein and the ability of hypoxic T47D cell-conditioned media to promote tumor angiogenesis in vitro. Under hypoxic conditions, 1 (10 microM) blocked the induction of HIF-1alpha protein, the oxygen-regulated subunit that controls HIF-1 activity. Reactive oxygen species produced by mitochondrial complex III are believed to act as a signal of cellular hypoxia that leads to HIF-1alpha protein induction and activation. Further mechanistic studies revealed that 1 inhibits mitochondrial respiration at electron transport chain (ETC) complex I (NADH-ubiquinone oxidoreductase). Under hypoxic conditions, it is proposed that 1 may disrupt mitochondrial ROS-regulated HIF-1 activation and HIF-1 downstream target gene expression by inhibiting the transport or delivery of electrons to complex III.
Sigma-Aldrich
1,10-Phenanthroline, ≥99%