MilliporeSigma
  • A facile 1,3-dipolar cycloaddition of azomethine ylides to 2-arylidene-1,3-indanediones: synthesis of dispiro-oxindolylpyrrolothiazoles and their antimycobacterial evaluation.
A facile 1,3-dipolar cycloaddition of azomethine ylides to 2-arylidene-1,3-indanediones: synthesis of dispiro-oxindolylpyrrolothiazoles and their antimycobacterial evaluation.
Bioorganic & medicinal chemistry letters (2010-11-13)
Shanmugavel Uma Maheswari, Kamaraj Balamurugan, Subbu Perumal, Perumal Yogeeswari, Dharmarajan Sriram
ABSTRACT
A facile 1,3-dipolar cycloaddition of azomethine ylide generated in situ from the reaction of 1,3-thiazolane-4-carboxylic acid and isatin to 2-arylidene-1,3-indanediones furnished novel dispiro-oxindolylpyrrolothiazoles regio- and stereo-selectively in moderate to good yields (60-92%). In vitro antitubercular screening of 27 compounds against Mycobacterium tuberculosis H37Rv (MTB) disclosed that spiro[5.3']-5'-nitrooxindolespiro-[6.3″]-1H-inden-1″,3″(2H)-dione-7-(4-bromophenyl)tetrahydro-1H-pyrrolo[1,2-c][1,3]thiazole has the maximum potency with a minimum inhibitory concentration (MIC) of 1.4 μM against MTB, being 3.4 and 5.4 times more potent than ciprofloxacin and ethambutol, respectively.
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Supelco
Isoniazid, analytical standard, ≥99% (TLC)
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)