MilliporeSigma
  • Association of Hydrophobic Carboxyl-Terminal Dendrimers with Lymph Node-Resident Lymphocytes.
Association of Hydrophobic Carboxyl-Terminal Dendrimers with Lymph Node-Resident Lymphocytes.
Polymers (2020-07-08)
Yutaka Nishimoto, Misaki Nishio, Shu Nagashima, Kohei Nakajima, Takayuki Ohira, Shinya Nakai, Ikuhiko Nakase, Kei Higashikawa, Yuji Kuge, Akikazu Matsumoto, Mikako Ogawa, Chie Kojima
ABSTRACT
Delivery systems to lymph node-resident T cells around tumor tissues are essential for cancer immunotherapy, in order to boost the immune responses. We previously reported that anionic dendrimers, such as carboxyl-, sulfonyl-, and phosphate-terminal dendrimers, were efficiently accumulated in lymph nodes via the intradermal injection. Depending on the terminal structure, their cell association properties were different, and the carboxyl-terminal dendrimers did not associate with any immune cells majorly. In this study, we investigated the delivery of carboxyl-terminal dendrimers with different hydrophobicity to lymph node-resident lymphocytes. Four types of carboxyl-terminal dendrimers-succinylated (C) and 2-carboxy-cyclohexanoylated (Chex) dendrimers with and without phenylalanine (Phe)-were synthesized and named C-den, C-Phe-den, Chex-den, and Chex-Phe-den, respectively. Chex-Phe-den was well associated with lymphocytes, but others were not. Chex-Phe-den, intradermally injected at the footpads of mice, was accumulated in the lymph node, and was highly associated with the lymphocytes, including T cells. Our results suggest that Chex-Phe-den has the potential for delivery to the lymph node-resident T cells, without any specific T cell-targeted ligands.
Sigma-Aldrich
N-Boc-ethylenediamine, ≥98.0% (NT)
Sigma-Aldrich
4-(4,6-Dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride, 95% (HPLC)